Tisk…

1-Bromobutane-2,3-dione 97%

Dodavatel: Apollo Scientific
Varování

OR46042-5G OR46042-1G
APOSOR46042-5GEA 5920 CZK
APOSOR46042-5G APOSOR46042-1G
1-Bromobutane-2,3-dione 97%
1-Bromobutane-2,3-dione
Vzorec: C₄H₅BrO₂
Storage Temperature: Refrigerator
CAS číslo: 5308-51-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR