Tisk…
ASTM Ubbelohde viscometers
Dodavatel: SI Analytics
285401152 285401169 285401177 285401185 285401193 285401255 285401263 285401271 285401288 285401296 285401309
SCOI285401152EA 14650 CZK
SCOI285401152 SCOI285401169 SCOI285401177 SCOI285401185 SCOI285401193 SCOI285401255 SCOI285401263 SCOI285401271 SCOI285401288 SCOI285401296 SCOI285401309
ASTM Ubbelohde viscometers
Viskozimetry Viskozimetry kapilární
Viscometers with suspended ball level for determination of relative viscosity of liquids with Newtonian flow behaviour.
Recommended minimum flow-through time is 200 seconds.
Certifikace: DIN 51 562 Part 1, ISO/DIN 3105 (BS-IP-SL).
ATSM calibrated for automatic measurements.