Tisk…
CRS GC Septa, Marathon Long-life
Dodavatel: Trajan Scientific and Medical
041907 041908 041910 041911 041913 041914 041915 239297 239397 239498 239597 239698 239779 239798 239888 239587 239778
SGEA041907EA 1970 CZK
SGEA041907 SGEA041908 SGEA041910 SGEA041911 SGEA041913 SGEA041914 SGEA041915 SGEA239297 SGEA239397 SGEA239498 SGEA239597 SGEA239698 SGEA239779 SGEA239798 SGEA239888 SGEA239587 SGEA239778
CRS GC Septa, Marathon Long-life
Septum
Marathon long-life center guide GC septa are ideal for standard manual injection GC and GC/MS.
  • Reliabily seals against the carrier gas pressure in the inlet
  • Excellent durability and resealing
  • Good solvent and tear resistance
  • Does not contaminate or bleed material into the chromatographic system
  • Long life septa, pre-pierced for minimal coring
  • Typically achieves up to 400 injections without failure
These GC septa are made up of silicone and are capable of being pierced and resealing time after time without pieces of the septa being deposited in the GCMore Product Information