Tisk…
Micro-Ostwald viscometers
Dodavatel: SI Analytics
285404211 285404425 285404203 285404228 285404306 285404322 285404339 285404347 285404314 285404236 285404244 285404409 285404417 285404433 285404441
SCOI285404211EA 11370 CZK
SCOI285404211 SCOI285404425 SCOI285404203 SCOI285404228 SCOI285404306 SCOI285404322 SCOI285404339 SCOI285404347 SCOI285404314 SCOI285404236 SCOI285404244 SCOI285404409 SCOI285404417 SCOI285404433 SCOI285404441
Micro-Ostwald viscometers
Viskozimetry Viskozimetry kapilární
These Micro-Ostwald viscometers are available as calibrated for either manual or automatic measurements and uncalibrated models.
Suitable for measurements of small liquid quantities with extreme formation of foam.