Tisk…
Reverse flow Cannon-Fenske viscometers for manual measurements
Dodavatel: SI Analytics
285403067 285403018 285403034 285403001 285403026 285403042 285403059 285403075 285403083 285403091 285403104 285403112
SCOI285403067EA 11860 CZK
SCOI285403067 SCOI285403018 SCOI285403034 SCOI285403001 SCOI285403026 SCOI285403042 SCOI285403059 SCOI285403075 SCOI285403083 SCOI285403091 SCOI285403104 SCOI285403112
Reverse flow Cannon-Fenske viscometers for manual measurements
Viskozimetry Viskozimetry kapilární
Reverse flow viscometers calibrated with constant.
Suitable for opaque liquids manual measurements.