Tisk…
Human sera
Dodavatel: Biowest
S419H-100 S4200-100 S4190-100 S419G-100 S420H-100
S419H-100EA 12200 CZK
S419H-100 S4200-100 S4190-100 S419G-100 S420H-100
Human sera
Séra
Human serum 'off-the-clot' is processed from human blood that has been coagulation-induced. It is collected from volunteer donors.
Mainly sourced from France, Germany, Canada or USA, each batch is rigorously controlled and screened for Hepatitis B (HBS), Hepatitis C (HCV) and HIV Types 1 and 2 (HIV 1/2).
Important note: Products of human origin should be considered potentially infectious and handled accordingly.
Informace pro objednávání: Special treatments are available on request. Please contact your local Avantor Sales Office for more information.