Tisk…
Foetal bovine serum (FBS), South American origin
Dodavatel: Biowest
S1810-100 S1810-500 S181D-100 S181D-500 S181F-100 S181G-100 S181G-500 S181H-100 S181H-500 S181L-100 S181R-100 S181R-500 S1860-500 S181S-500 S181T-100 S181T-500 S1860-100 S181F-500 S181S-100 S181L-500 S186G-500 S186G-100 S181M-050 S181M-100 S181M-500 S181I-500 S181W-500
S1810-100EA 2470 CZK
S1810-100 S1810-500 S181D-100 S181D-500 S181F-100 S181G-100 S181G-500 S181H-100 S181H-500 S181L-100 S181R-100 S181R-500 S1860-500 S181S-500 S181T-100 S181T-500 S1860-100 S181F-500 S181S-100 S181L-500 S186G-500 S186G-100 S181M-050 S181M-100 S181M-500 S181I-500 S181W-500
Foetal bovine serum (FBS), South American origin
Séra
Sterile filtered foetal bovine serum (FBS) derived from clotted whole blood aseptically collected from foetus via cardiac puncture. Each manufactured batch is rigorously controlled, from the collection of serum through to final packaging.
  • South American origin
  • High quality serum, rigorously tested
  • Full traceability and security
Certifikace: Biowest is a certified ISO 9001:2008 company. Registered by the French Ministry of Agriculture (EC regulation No. 1069/2009) under agreement no. FR 49.231.001 for the production of animal sera.