Tisk…
Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer
Dodavatel: Thermo Fisher Scientific
17908 17909
PIER17908EA 3740 CZK
PIER17908 PIER17909
Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer
Pufry Pufry extrakční
Phenol-chloroform, is phenol supplied premixed with chloroform:isoamyl alcohol (25:24:1) and accompanying bottle of buffer to equilibrate at either pH 6,7 or pH 8,0.
  • Offers excellent, nuclease-free performance
  • Ideal for lower pH DNA extractions and for most DNA applications