Tisk…
Septa for 13-425 screw thread caps
Dodavatel: THERMO NATIONAL SCIENTIFIC
C4015-60 C4015-40
NSCAC4015-60EA 1800 CZK
NSCAC4015-60 NSCAC4015-40
Septa for 13-425 screw thread caps
Septum
These septa are suitable for use with 13-425 threaded caps or 13 mm snap/crimp caps.