Tisk…
Immersion circulator, Clifton NE4-T
Dodavatel: NICKEL ELECTRO
NE4-T
NICKNE4-TEA 37770 CZK
NICKNE4-T
Immersion circulator, Clifton NE4-T
Termostaty oběhové Otevřené cirkulační systémy Ovládácí jednotky termostatu
Durable stainless steel construction, painted surfaces are powder painted with tough anti-bacterial finish with good chemical resistance. Easy to use analogue controls that wipe clean and have an anti-bacterial finish. Immersed components are corrosion resistant. Quiet running in use and providing good water temperature stability. Indication of heating and low liquid levels.
  • Thermostatic control, easy to use dial setting, ambient 5 °C to 99 °C
  • Powerful stirring within the tank for good water circulation and temperature stability
  • Controls with an
More Product Information