Tisk…
Immersion Circulators, ICC 150 lite pro 9 c
Dodavatel: IKA
10013233
IKAA10013233EA 69860 CZK
IKAA10013233
Immersion Circulators, ICC 150 lite pro 9 c
Termostaty oběhové Otevřené cirkulační systémy Ovládácí jednotky termostatu
The compact immersion circulators of the lite series are powerful and of high quality, with a focus on essential basic functions, excellent performance and efficient operation.
  • Max. temperature in stainless steel bath (pro): 150 °C
ICC 150 lite is suitable for tempering liquids up to 150 °C. The integrated pressure/ suction pump allows ICC 150 lite to be used for internal and external applications.
To make configuration of your temperature control system as convenient as possible, IKA offers 'ready-to-connect' packages with all the necessaryMore Product Information