Tisk…
Baths and circulators, stainless steel
Dodavatel: HUBER
2038.0021.99 2057.0001.01 2056.0001.01 2058.0001.01 2059.0001.01 2060.0001.01
461-0557EA 36770 CZK
461-0557 HUBE2057.0001.01 HUBE2056.0001.01 HUBE2058.0001.01 HUBE2059.0001.01 HUBE2060.0001.01
Baths and circulators, stainless steel
Termostaty oběhové Otevřené cirkulační systémy Lázně s odnímatelným oběhovým čerpadlem
Circulating bath thermostats with baths made from stainless steel are simple to use and suitable for many applications such as: sample temperature control, analysis, material testing or checking food. The units offer high temperature stability and are fitted with over-temperature and low level protection suitable for use with flammable fluids. A powerful pressure/suction pump ensures optimum circulation and temperature homogenity in the bath and also allows temperature control of external applications using a pump adapter (accessory). There is a range of optional accessories includingMore Product Information