Tisk…
Kits: in search of the sickle cell gene by southern blot, Ready-to-Load™
Dodavatel: EDVOTEK
315
EDVO315EA 5110 CZK
EDVO315
Kits: in search of the sickle cell gene by southern blot, Ready-to-Load™
Materiály výukové Materiál pro výuku biologie Aktivity pro výuku genetiky
Southern blotting is an important technique used widely in clinical genetics and research. By transferring DNA from an agarose gel onto a membrane the method allows the precise analysis and identification of DNA bands on a gel. Using this kit, students use Southern blotting to find a point mutation in the haemoglobin gene indicating Sickle Cell Anaemia.
  • For five lab groups
  • Electrophoresis, 45 minutes; blotting, overnight; staining and destaining, 10 minutes
Also required: Electrophoresis tank, power supply, water bath and 80 °C incubator.