Tisk…
Experimentation kits, diversity of fish proteins
Dodavatel: EDVOTEK
253
EDVO253EA 2260 CZK
EDVO253
Experimentation kits, diversity of fish proteins
Materiály výukové Materiál pro výuku biologie Aktivity pro výuku genetiky
In this experiment, students study the diversity of fish using pre-stained, lyophilised proteins. Total protein from perch, walleye and salmon is extracted and pre-stained using an indicator dye. Each fish protein sample has a characteristic banding pattern when separated by denaturing SDS-polyacrylamide gel electrophoresis, which can be used to identify the specific species.
  • For 6 lab groups (sharing 3 gels)
  • Electrophoresis 60 minutes, staining 20 minutes, destaining 2 hours
Also required: 3 polyacrylamide gels (12%), vertical gel electrophoresisMore Product Information