Tisk…
Experimentation kits, southern blot analysis
Dodavatel: EDVOTEK
207
EDVO207EA 3970 CZK
EDVO207
Experimentation kits, southern blot analysis
Materiály výukové Materiál pro výuku biologie Aktivity pro výuku fyziologie a zdraví
This experiment introduces students to Southern blotting as a tool for DNA fingerprinting in a hypothetical paternity determination. DNA fragments are first separated by agarose gel electrophoresis, then transferred to a nylon membrane and finally visualised by staining.
  • For 5 lab groups
  • Electrophoresis 45 minutes, blotting overnight, staining and destaining 10 minutes
Also required: Electrophoresis tank and power supply, 65 °C water batch, DNA visualisatoin system, staining net and tray, automatic micropipettes, lab glassware, microwave oven,More Product Information