Tisk…
Kits: mapping of restriction sites on plasmid DNA, Ready-to-Load™
Dodavatel: EDVOTEK
105 105-C
EDVO105EA 2830 CZK
EDVO105 EDVO105-C
Kits: mapping of restriction sites on plasmid DNA, Ready-to-Load™
Materiály výukové Materiál pro výuku biologie Aktivity pro výuku genetiky
DNA mapping is a common procedure used to determine the location of genes. In this experiment, DNA markers and pre-digested plasmid DNA fragments are mapped using agarose gel electrophoresis.
  • For six lab groups
Also required: Electrophoresis tank, power supply, micropipettes (5 to 50 µl adjustable or 40 µl fixed volume) with tips, and a white light box is recommended.
Informace o dodávce: Kit includes instructions, Ready-to-Load™ DNA samples, agarose powder, practice gel loading solution, electrophoresis buffer, InstaStain® DNA stain, 1 mlMore Product Information