Tisk…
Electrodes, Genetrodes™
Dodavatel: BTX
45-0113 45-0114 45-0115 45-0116 45-0117 45-0161 45-0162 45-0160 45-0163 45-0164
732-0010EA 16790 CZK
732-0010 732-0011 732-0012 732-0013 732-0014 BTXI45-0161 BTXI45-0162 732-0933 BTXI45-0163 BTXI45-0164
Electrodes, Genetrodes™
Elektrody Elektrody elektroporační
Genetrodes™ are paired, reusable, needle style electrodes that are ideal for in vivo and in ovo electroporation applications, including drug and gene delivery.
  • Five models to suit the size and shape of the target electroporation area
  • Each model consists of a pair of electrodes configured as either straight or bent L-shaped electrodes with gold tips
  • The electrodes are placed into tissue following injection of the molecule of interest, and an electroporation pulse is delivered using an electroporation generator
  • The electric field introduced by the Genetrodes™
More Product Information