Tisk…
VWR®, Septa ND20
Dodavatel: VWR Collection
548-0568AEA 18900 CZK
548-0568A 548-0571A 548-0570A 548-1979A 548-0565A 548-0797A 548-0569A 548-1833A 548-0567A 548-0573A 548-1059A 548-1484A 548-1456A 548-0572A 548-0807A 548-0869A
VWR®, Septa ND20
Septum
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.