Tisk…
Cooling and heating thermostats, stainless steel, with natural refrigerant, Ministats
Dodavatel: HUBER
2015.0007.01 2016.0006.01 2016.0014.04 2014.0006.01 2014.0011.01 2015.0005.01 2016.0005.01
461-0739EA 204970 CZK
461-0739 461-0740 HUBE2016.0014.04 461-1003 461-0803 461-0744 461-0745
Cooling and heating thermostats, stainless steel, with natural refrigerant, Ministats
Termostaty oběhové Otevřené cirkulační systémy Termostaty cirkulační pro ohřev/chlazení
Their compact dimensions enable Ministats® to be placed in the smallest of spaces, for example in a laboratory fume cupboard or inside technical plant. Despite their minimal size the systems have a wide variety of features and offer performance levels suitable for temperature control of photometers, refractometers, viscometers, distillation apparatus, reaction vessels and mini-plants. They are mainly used for external applications, however the bath opening allows objects to be thermoregulated directly in a thermostat bath. A powerful pressure/suction pump with an infinitely controlledMore Product Information