Tisk…
u-fit ft Chemical Protection Gloves, 60166 Series
Dodavatel: UVEX
6016606 6016607 6016608 6016609 6016610
UVEX6016606EA 500 CZK
UVEX6016606 UVEX6016607 UVEX6016608 UVEX6016609 UVEX6016610
u-fit ft Chemical Protection Gloves, 60166 Series
Rukavice
The uvex u-fit ft is characterised by its excellent mechanical strength and also offers reliable protection against oil and dirt for light assembly activities.
  • Outstanding tactile feel
  • Very good fit
  • Extremely high flexibility
Thin and reliable NBR disposable safety gloves (0,10 mm).