Tisk…
athletic D5 XP Cut Protection Gloves
Dodavatel: UVEX
6003009 6003010 6003011 6003006 6003007 6003008
UVEX6003009EA 3800 CZK
UVEX6003009 UVEX6003010 UVEX6003011 UVEX6003006 UVEX6003007 UVEX6003008
athletic D5 XP Cut Protection Gloves
Rukavice
The athletic D5 XP cut protection gloves boast very high level D cut protection and a reinforced thumb joint. The comfortable fit and knitted cuff ensure that wearers can work flexibly while enjoying high levels of mechanical protection.
  • Cut protection glove with additional thumb reinforcement
  • Flexible safety gloves with excellent fit due to unique uvex ergonomic liner
  • Knitted cuff
Certified in line with OEKO-TEX® Standard 100, the uvex athletic range of safety gloves has a particularly comfortable slim fit. The lightweight gloves with a highly breathable microfoam coating ensure flexible and sensitive working with a high degree of mechanical protection. Outer material is made from HPPE, steel, and polyamide/elastane, with a micro-NBR foam coating on palm and fingertips.