Tisk…
Safety Gloves phynomic wet plus, 60061
Dodavatel: UVEX
6006106 6006112
UVEX6006106EA 1990 CZK
UVEX6006106 UVEX6006112
Safety Gloves phynomic wet plus, 60061
Rukavice
The phynomic series is ideal for all types of work where a high level of feeling is required.
  • Good grip in dry, wet, or oily conditions
  • Very high abrasion resistance
  • Excellent skin compatibility and free from harmful substances
These safety gloves fit like a second skin and are particularly light and flexible, without making the hands sweat. The anthracite-coloured aqua-polymer foam coating offers outstanding protection against humidity. An aqua-polymer foam coating on palm, fingertips, and parts of the back of the hand serves as a water-repellent barrier for outdoor use.