Tisk…
u-chem 3100 Safety Gloves, 60968 Series
Dodavatel: UVEX
6096810 6096808 6096809
UVEX6096810EA 3900 CZK
UVEX6096810 UVEX6096808 UVEX6096809
u-chem 3100 Safety Gloves, 60968 Series
Rukavice
The u-chem 3100 is a full chemical protection glove that also offers good mechanical protection and is ideal for use in a wide range of applications.
  • Very high abrasion resistance
  • Good chemical resistance
  • Gauntlet design
Chemical protection gauntlet glove with excellent grip, very good mechanical protection, broad chemical resistance, and excellent grip in wet and oily conditions.