Tisk…

Anti-Rab11A Rabbit Monoclonal Antibody [clone: EPR28202-94]

Dodavatel: Abcam
AB316151-20UL AB316151-100UL AB316151-1ML
ABCAAB316151-20ULEA 6950 CZK
ABCAAB316151-20UL ABCAAB316151-100UL ABCAAB316151-1ML
Anti-Rab11A Rabbit Monoclonal Antibody [clone: EPR28202-94]
Protilátky
Rabbit monoclonal [EPR28202-94] to Rab11A.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Rab11A
Clonality: Monoclonal
Clone: EPR28202-94
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR