Tisk…

Anti-MYL9+MYL12A+MYL12B (phospho S20) Rabbit Monoclonal Antibody [clone: EPR27203-65]

Dodavatel: Abcam
AB316751-1MG AB316751-100UG
ABCAAB316751-1MGEA 260960 CZK
ABCAAB316751-1MG ABCAAB316751-100UG
Anti-MYL9+MYL12A+MYL12B (phospho S20) Rabbit Monoclonal Antibody [clone: EPR27203-65]
Protilátky
Rabbit monoclonal [EPR27203-65] to MYL9 (phospho S20) + MYL12A (phospho S19) + MYL12B (phospho S20) - BSA and Azide free.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: MYL9+MYL12A+MYL12B
Clonality: Monoclonal
Clone: EPR27203-65
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR