Tisk…

Anti-ErbB2/HER2 (phospho Y1221 + Y1222) Rabbit Monoclonal Antibody [clone: EPR28391-67]

Dodavatel: Abcam
AB316758-20UL AB316758-100UL AB316758-1ML
ABCAAB316758-20ULEA 6950 CZK
ABCAAB316758-20UL ABCAAB316758-100UL ABCAAB316758-1ML
Anti-ErbB2/HER2 (phospho Y1221 + Y1222) Rabbit Monoclonal Antibody [clone: EPR28391-67]
Protilátky
Rabbit monoclonal [EPR28391-67] to ErbB2 / HER2 (phospho Y1221 + Y1222).

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ErbB2 / HER2
Clonality: Monoclonal
Clone: EPR28391-67
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR