Tisk…

Anti-CTGF Rabbit Recombinant Multiclonal Antibody [clone: RM1199]

Dodavatel: Abcam
AB318149-100UG AB318149-1MG
ABCAAB318149-100UGEA 29040 CZK
ABCAAB318149-100UG ABCAAB318149-1MG
Anti-CTGF Rabbit Recombinant Multiclonal Antibody [clone: RM1199]
Protilátky
Rabbit recombinant multiclonal [RM1199] to CTGF - BSA and Azide free.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CTGF
Clonality: Recombinant Multiclonal
Clone: RM1199
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR