Tisk…

Anti-delta Sarcoglycan Rabbit Monoclonal Antibody [clone: EPR8706] (Alexa Fluor® 647)

Dodavatel: Abcam
AB319130-10UL AB319130-100UL
ABCAAB319130-10ULEA 6770 CZK
ABCAAB319130-10UL ABCAAB319130-100UL
Anti-delta Sarcoglycan Rabbit Monoclonal Antibody [clone: EPR8706] (Alexa Fluor® 647)
Protilátky
Alexa Fluor® 647 Rabbit monoclonal [EPR8706] to delta Sarcoglycan.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: delta Sarcoglycan
Clonality: Monoclonal
Clone: EPR8706
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR