Tisk…

Anti-AAV9 Mouse Monoclonal Antibody [clone: HL2368]

Dodavatel: Abcam
AB315819-1MG AB315819-100UG
ABCAAB315819-1MGEA 260960 CZK
ABCAAB315819-1MG ABCAAB315819-100UG
Anti-AAV9 Mouse Monoclonal Antibody [clone: HL2368]
Protilátky
Mouse monoclonal [HL2368] to AAV9 - BSA and Azide free.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: AAV9
Clonality: Monoclonal
Clone: HL2368
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: AAV-9
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR