Tisk…

Anti-AAV9 Mouse Monoclonal Antibody [clone: HL2370]

Dodavatel: Abcam
AB291244-1MG AB291244-100UG
ABCAAB291244-1MGEA 260960 CZK
ABCAAB291244-1MG ABCAAB291244-100UG
Anti-AAV9 Mouse Monoclonal Antibody [clone: HL2370]
Protilátky
Mouse monoclonal [HL2370] to AAV9 - BSA and Azide free.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: AAV9
Clonality: Monoclonal
Clone: HL2370
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Adeno-associated virus 9
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR