Tisk…

Anti-Lamin A + Lamin C Rabbit Recombinant Multiclonal Antibody [clone: RM1093]

Dodavatel: Abcam
AB315839-1MG AB315839-100UG
ABCAAB315839-1MGEA 260960 CZK
ABCAAB315839-1MG ABCAAB315839-100UG
Anti-Lamin A + Lamin C Rabbit Recombinant Multiclonal Antibody [clone: RM1093]
Protilátky
Rabbit recombinant multiclonal [RM1093] to Lamin A + Lamin C - BSA and Azide free.

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Lamin A + Lamin C
Clonality: Recombinant Multiclonal
Clone: RM1093
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR