Tisk…

Anti-SPTLC1 Rabbit Monoclonal Antibody [clone: EPR26226-221]

Dodavatel: Abcam
AB315158-20UL AB315158-100UL AB315158-1ML
ABCAAB315158-20ULEA 6950 CZK
ABCAAB315158-20UL ABCAAB315158-100UL ABCAAB315158-1ML
Anti-SPTLC1 Rabbit Monoclonal Antibody [clone: EPR26226-221]
Protilátky
Rabbit monoclonal [EPR26226-221] to SPTLC1.

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SPTLC1
Clonality: Monoclonal
Clone: EPR26226-221
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR