Tisk…

Anti-MCM2 Rabbit Recombinant Multiclonal Antibody [clone: RM1146]

Dodavatel: Abcam
AB317753-100UG AB317753-1MG
ABCAAB317753-100UGEA 29040 CZK
ABCAAB317753-100UG ABCAAB317753-1MG
Anti-MCM2 Rabbit Recombinant Multiclonal Antibody [clone: RM1146]
Protilátky
Rabbit recombinant multiclonal [RM1146] to MCM2 - BSA and Azide free.

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MCM2
Clonality: Recombinant Multiclonal
Clone: RM1146
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR