Tisk…
Anti-Adenosine Receptor A2a Rabbit Monoclonal Antibody [clone: EPR23731-50]
Dodavatel: Abcam
AB260032-100UL AB260032-1ML
ABCAAB260032-100ULEA 19950 CZK
ABCAAB260032-100UL ABCAAB260032-1ML
Anti-Adenosine Receptor A2a Rabbit Monoclonal Antibody [clone: EPR23731-50]
Protilátky
Rabbit monoclonal [EPR23731-50] to Adenosine Receptor A2a.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Adenosine Receptor A2a
Clonality: Monoclonal
Clone: EPR23731-50
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat