Tisk…

Anti-Mu Opioid Receptor Rabbit Monoclonal Antibody [clone: EPR26293-61]

Dodavatel: Abcam
AB302512-100 AB302512-20UL AB302512-1ML
ABCAAB302512-100EA 19240 CZK
ABCAAB302512-100 ABCAAB302512-20UL ABCAAB302512-1ML
Anti-Mu Opioid Receptor Rabbit Monoclonal Antibody [clone: EPR26293-61]
Protilátky
Rabbit monoclonal [EPR26293-61] to Mu Opioid Receptor.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Mu Opioid Receptor
Clonality: Monoclonal
Clone: EPR26293-61
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR