Tisk…

Anti-OCLN Rabbit Monoclonal Antibody [clone: EPR20992]

Dodavatel: Abcam
AB224526-100 AB224526-1
ABCAAB224526-100EA 29040 CZK
ABCAAB224526-100 ABCAAB224526-1
Anti-OCLN Rabbit Monoclonal Antibody [clone: EPR20992]
Protilátky
Rabbit monoclonal [EPR20992] to Occludin - BSA and Azide free.

Type: Primary
Antigen: OCLN
Clonality: Monoclonal
Clone: EPR20992
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Dog
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR