Tisk…
Anti-ATF2 (phospho T71) Rabbit Monoclonal Antibody [clone: EPR268-111]
Dodavatel: Abcam
AB308570-100UG AB308570-1MG
ABCAAB308570-100UGEA 26190 CZK
ABCAAB308570-100UG ABCAAB308570-1MG
Anti-ATF2 (phospho T71) Rabbit Monoclonal Antibody [clone: EPR268-111]
Protilátky
Rabbit monoclonal [EPR268-111] to ATF2 (phospho T71) - BSA and Azide free (Detector).
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: ATF2
Clonality: Monoclonal
Clone: EPR268-111
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human