Tisk…

Anti-xCT Rat Monoclonal Antibody [clone: 3A12]

Dodavatel: Abcam
AB300667-100 AB300667-20UG AB300667-1MG
ABCAAB300667-100EA 20130 CZK
ABCAAB300667-100 ABCAAB300667-20UG ABCAAB300667-1MG
Anti-xCT Rat Monoclonal Antibody [clone: 3A12]
Protilátky
Rat monoclonal [3A12] to xCT.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: xCT
Clonality: Monoclonal
Clone: 3A12
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR