Tisk…

Anti-Metabotropic Glutamate Receptor 5 Rabbit Monoclonal Antibody [clone: EPR2425Y]

Dodavatel: Abcam
AB219374-100 AB219374-1
ABCAAB219374-100EA 29040 CZK
ABCAAB219374-100 ABCAAB219374-1
Anti-Metabotropic Glutamate Receptor 5 Rabbit Monoclonal Antibody [clone: EPR2425Y]
Protilátky
Rabbit monoclonal [EPR2425Y] to Metabotropic Glutamate Receptor 5 - Low endotoxin, Azide free.

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Metabotropic Glutamate Receptor 5
Clonality: Monoclonal
Clone: EPR2425Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR