Tisk…

Anti-Delta Opioid Receptor Rabbit Monoclonal Antibody [clone: EPR5029(2)]

Dodavatel: Abcam
AB176324-100 AB176324-1ML
ABCAAB176324-100EA 17460 CZK
ABCAAB176324-100 ABCAAB176324-1ML
Anti-Delta Opioid Receptor Rabbit Monoclonal Antibody [clone: EPR5029(2)]
Protilátky
Rabbit monoclonal [EPR5029(2)] to Delta Opioid Receptor.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Delta Opioid Receptor
Clonality: Monoclonal
Clone: EPR5029(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR