Tisk…

Anti-UFM1 Rabbit Monoclonal Antibody [clone: EPR4264(2)]

Dodavatel: Abcam
AB232570-100 AB232570-1
ABCAAB232570-100EA 29040 CZK
ABCAAB232570-100 ABCAAB232570-1
Anti-UFM1 Rabbit Monoclonal Antibody [clone: EPR4264(2)]
Protilátky
Rabbit monoclonal [EPR4264(2)] to UFM1 - BSA and Azide free.

Type: Primary
Antigen: UFM1
Clonality: Monoclonal
Clone: EPR4264(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR