Tisk…

Anti-OCT6 Rabbit Monoclonal Antibody [clone: EPR24057-94]

Dodavatel: Abcam
AB259952-100 AB259952-20UL AB259952-1ML
ABCAAB259952-100EA 20310 CZK
ABCAAB259952-100 ABCAAB259952-20UL ABCAAB259952-1ML
Anti-OCT6 Rabbit Monoclonal Antibody [clone: EPR24057-94]
Protilátky
Rabbit monoclonal [EPR24057-94] to Oct6.

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: OCT6
Clonality: Monoclonal
Clone: EPR24057-94
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR