Tisk…

Anti-HLA-DR Mouse Monoclonal Antibody [clone: TAL 1B5]

Dodavatel: Abcam
AB176408-100 AB176408-1
ABCAAB176408-100EA 0 CZK
ABCAAB176408-100 ABCAAB176408-1
Anti-HLA-DR Mouse Monoclonal Antibody [clone: TAL 1B5]
Protilátky
Mouse monoclonal [TAL 1B5] to HLA-DR - BSA and Azide free.

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: TAL 1B5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR