Tisk…

Anti-Cyclin T1 Rabbit Monoclonal Antibody [clone: EPR17982]

Dodavatel: Abcam
AB184703-100 AB184703-1ML
ABCAAB184703-100EA 20130 CZK
ABCAAB184703-100 ABCAAB184703-1ML
Anti-Cyclin T1 Rabbit Monoclonal Antibody [clone: EPR17982]
Protilátky
Rabbit monoclonal [EPR17982] to Cyclin T1.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cyclin T1
Clonality: Monoclonal
Clone: EPR17982
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR