Tisk…

Anti-GABA Rabbit Monoclonal Antibody [clone: EPR24433-9]

Dodavatel: Abcam
AB308437-100 AB308437-20UL AB308437-1ML
ABCAAB308437-100EA 19780 CZK
ABCAAB308437-100 ABCAAB308437-20UL ABCAAB308437-1ML
Anti-GABA Rabbit Monoclonal Antibody [clone: EPR24433-9]
Protilátky
Rabbit monoclonal [EPR24433-9] to GABA.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: GABA
Clonality: Monoclonal
Clone: EPR24433-9
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR