Tisk…

Anti-TVA Rat Monoclonal Antibody [clone: PICA187E]

Dodavatel: Abcam
AB271302-1 AB271302-100
ABCAAB271302-1EA 260960 CZK
ABCAAB271302-1 ABCAAB271302-100
Anti-TVA Rat Monoclonal Antibody [clone: PICA187E]
Protilátky
Rat monoclonal [PICA187E] to TVA - BSA and Azide free.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TVA
Clonality: Monoclonal
Clone: PICA187E
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Bird
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR