Tisk…

Anti-CD3 epsilon Rabbit Monoclonal Antibody [clone: EP449E]

Dodavatel: Abcam
AB213608-100 AB213608-1
ABCAAB213608-100EA 29040 CZK
ABCAAB213608-100 ABCAAB213608-1
Anti-CD3 epsilon Rabbit Monoclonal Antibody [clone: EP449E]
Protilátky
Rabbit monoclonal [EP449E] to CD3 epsilon - Low endotoxin, Azide free.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD3 epsilon
Clonality: Monoclonal
Clone: EP449E
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR