Tisk…
Anti-beta Catenin Rabbit Monoclonal Antibody [clone: EPR26155-110-1]
Dodavatel: Abcam
AB305261-100 AB305261-20UL AB305261-1ML
ABCAAB305261-100EA 13540 CZK
ABCAAB305261-100 ABCAAB305261-20UL ABCAAB305261-1ML
Anti-beta Catenin Rabbit Monoclonal Antibody [clone: EPR26155-110-1]
Protilátky
Rabbit monoclonal [EPR26155-110-1] to beta Catenin.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: beta Catenin
Clonality: Monoclonal
Clone: EPR26155-110-1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat