Tisk…

Anti-STARD4 Rabbit Monoclonal Antibody [clone: EPR17847]

Dodavatel: Abcam
AB251308-100 AB251308-1
ABCAAB251308-100EA 29040 CZK
ABCAAB251308-100 ABCAAB251308-1
Anti-STARD4 Rabbit Monoclonal Antibody [clone: EPR17847]
Protilátky
Rabbit monoclonal [EPR17847] to STARD4 - BSA and Azide free.

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: STARD4
Clonality: Monoclonal
Clone: EPR17847
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR