Tisk…

Anti-p53 (phospho S20) Rabbit Monoclonal Antibody [clone: EPR2156(2)]

Dodavatel: Abcam
AB157454-100 AB157454-20UL AB157454-1ML
ABCAAB157454-100EA 13540 CZK
ABCAAB157454-100 ABCAAB157454-20UL ABCAAB157454-1ML
Anti-p53 (phospho S20) Rabbit Monoclonal Antibody [clone: EPR2156(2)]
Protilátky
Rabbit monoclonal [EPR2156(2)] to p53 (phospho S20).

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: EPR2156(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR